Learn More
Guaiacol, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 120192500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Guaiacol | |
27.0°C to 29.0°C | |
1.1200g/mL | |
82°C | |
99% min. (GC) | |
C7H8O2 | |
2-(CH3O)C6H4OH | |
06,768 | |
15,4589 | |
Solubility in water: 17g/L (15°C). Other solubilities: miscible with glacial acetic acid, miscible with alcohol, chloroform and ether, slightly soluble in petroleum ether | |
COC1=CC=CC=C1O | |
124.14 | |
CHEBI:28591 | |
99+% |
90-05-1 | |
Colorless to Yellow | |
205.0°C | |
Authentic | |
Glass bottle | |
1.5430 to 1.5450 | |
250 g | |
1.12 | |
guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid | |
LHGVFZTZFXWLCP-UHFFFAOYSA-N | |
2-methoxyphenol | |
460 | |
124.14 | |
Liquid After Melting |
Chemical Identifiers
90-05-1 | |
124.14 | |
guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid | |
CHEBI:28591 | |
COC1=CC=CC=C1O |
C7H8O2 | |
LHGVFZTZFXWLCP-UHFFFAOYSA-N | |
460 | |
2-methoxyphenol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Warning
EINECSNumber : 201-964-7
RUO – Research Use Only