missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Guaiacol, 99+%, Thermo Scientific Chemicals

$82.89 - $183.50

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
PubChem CID 460
ChEBI CHEBI:28591
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120192500
SDS View Documents
Thermo Scientific Chemicals
120192500
250 g Glass bottle
Each for $82.89
 
AC120190010
SDS View Documents
Thermo Scientific Chemicals
120190010
1 kg Glass bottle
Each for $183.50
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90-05-1
124.14
guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
CHEBI:28591
COC1=CC=CC=C1O
C7H8O2
LHGVFZTZFXWLCP-UHFFFAOYSA-N
460
2-methoxyphenol

Specifications

90-05-1
Colorless to Yellow
205.0°C
Authentic
Glass bottle
1.5430 to 1.5450
250 g
1.12
guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
LHGVFZTZFXWLCP-UHFFFAOYSA-N
2-methoxyphenol
460
124.14
Liquid After Melting
27.0°C to 29.0°C
1.1200g/mL
82°C
99% min. (GC)
C7H8O2
2-(CH3O)C6H4OH
06,768
15,4589
Solubility in water: 17g/L (15°C). Other solubilities: miscible with glacial acetic acid, miscible with alcohol, chloroform and ether, slightly soluble in petroleum ether
COC1=CC=CC=C1O
124.14
CHEBI:28591
99+%
Guaiacol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Warning

EINECSNumber : 201-964-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only