missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 364790025

 View more versions of this product

Catalog No. AC364790025

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Guanidine hydrochloride
50-01-1
178.0°C to 188.0°C
1.3450g/mL
99.5%
CH5N3·HCl
MFCD00013026
03,82
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine;hydrochloride
5742
95.53
Crystals
Without anticaking agent
(6M aq. soln.), (6 M aq. soln.) at 0nm, 0.01 max. at 280nm, 0.03 max. at 260nm
Colorless to White
conform to structure and standard
Plastic bottle
H2NC(=NH)NH2·HCl
2.5 kg
15,4597
Solubility in water: 2150g/L (20°C).
C(=N)(N)N.Cl
95.53
CHEBI:32735
99.5%

Chemical Identifiers

50-01-1
95.53
PJJJBBJSCAKJQF-UHFFFAOYSA-N
5742
guanidine;hydrochloride
CH5N3·HCl
MFCD00013026
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
CHEBI:32735
C(=N)(N)N.Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 200-002-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only