missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Guanidine hydrochloride, 99.5%, without anticaking agent, Thermo Scientific Chemicals

$128.50 - $4137.40

Chemical Identifiers

CAS 50-01-1
Molecular Formula CH5N3·HCl
Molecular Weight (g/mol) 95.53
MDL Number MFCD00013026
InChI Key PJJJBBJSCAKJQF-UHFFFAOYSA-N
Synonym guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PubChem CID 5742
ChEBI CHEBI:32735
IUPAC Name guanidine;hydrochloride
SMILES C(=N)(N)N.Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC364791000
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
364791000
100 g Plastic bottle
Each for $128.50
 
AC364795000
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
364795000
500 g Plastic bottle
Each for $322.72
 
AC364790025
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
364790025
2.5 kg Plastic bottle
Each for $1,401.60
 
AC364790100
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
364790100
10 kg Plastic Drum
Each for $4,137.40
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-01-1
95.53
PJJJBBJSCAKJQF-UHFFFAOYSA-N
5742
guanidine;hydrochloride
CH5N3·HCl
MFCD00013026
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
CHEBI:32735
C(=N)(N)N.Cl

Specifications

50-01-1
178.0°C to 188.0°C
1.3450g/mL
99.5%
CH5N3·HCl
MFCD00013026
03,82
guanidine hydrochloride, guanidinium chloride, guanidine, monohydrochloride, aminoformamidine hydrochloride, aminomethanamidine hydrochloride, guanidine monohydrochloride, guanidine hcl, guanidine chloride, iminourea hydrochloride, carbamidine hydrochloride
PJJJBBJSCAKJQF-UHFFFAOYSA-N
guanidine;hydrochloride
5742
95.53
Crystals
(6M aq. soln.), (6 M aq. soln.) at 0nm, 0.01 max. at 280nm, 0.03 max. at 260nm
Colorless to White
conform to structure and standard
Plastic bottle
H2NC(=NH)NH2·HCl
100 g
15,4597
Solubility in water: 2150g/L (20°C).
C(=N)(N)N.Cl
95.53
CHEBI:32735
99.5%
Guanidine hydrochloride, Without anticaking agent
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 200-002-3

SDS
Documents

Documents

RUO – Research Use Only