Learn More
Guanidine nitrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L096430B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Guanidine nitrate | |
213°C to 216°C | |
4.9 | |
CH6N4O3 | |
1000 g | |
3596600 | |
guanidine nitrate, guanidinium nitrate, guanidine; nitric acid, guanidine, mononitrate, guanidine, nitrate, guanidine mononitrate, guanidine nitrate 1:1, guanidinenitrate, pubchem21372, acmc-209kpa | |
C(=N)(N)N.[N+](=O)(O)[O-] | |
122.084 | |
122.08 |
506-93-4 | |
1.44 | |
Odorless | |
MFCD00013028 | |
UN1467 | |
14,4562 | |
CNUNWZZSUJPAHX-UHFFFAOYSA-N | |
guanidine;nitric acid | |
10481 | |
98% |
Chemical Identifiers
506-93-4 | |
122.084 | |
CNUNWZZSUJPAHX-UHFFFAOYSA-N | |
10481 | |
C(=N)(N)N.[N+](=O)(O)[O-] |
CH6N4O3 | |
MFCD00013028 | |
guanidine nitrate, guanidinium nitrate, guanidine; nitric acid, guanidine, mononitrate, guanidine, nitrate, guanidine mononitrate, guanidine nitrate 1:1, guanidinenitrate, pubchem21372, acmc-209kpa | |
guanidine;nitric acid |
Safety and Handling
GHS H Statement
H272-H302-H315-H319
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P210-P220-P221-P261-P264b-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P370+P380+P375-P372-P380-P501c
H272-H302-H315-H319-H350-X
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: GUANIDINE NITRATE
EINECSNumber : 208-060-1
RTECSNumber : MF4350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only