missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Guanidine nitrate, 98%, Thermo Scientific Chemicals

$59.81 - $165.49

Chemical Identifiers

CAS 506-93-4
Molecular Formula CH6N4O3
Molecular Weight (g/mol) 122.084
MDL Number MFCD00013028
InChI Key CNUNWZZSUJPAHX-UHFFFAOYSA-N
Synonym guanidine nitrate, guanidinium nitrate, guanidine; nitric acid, guanidine, mononitrate, guanidine, nitrate, guanidine mononitrate, guanidine nitrate 1:1, guanidinenitrate, pubchem21372, acmc-209kpa
PubChem CID 10481
IUPAC Name guanidine;nitric acid
SMILES C(=N)(N)N.[N+](=O)(O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0964330
View Documents
Thermo Scientific Chemicals
L0964330
250 g
Each for $59.81
 
AAL096430B
View Documents
Thermo Scientific Chemicals
L096430B
1000 g
Each for $165.49
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

506-93-4
122.084
CNUNWZZSUJPAHX-UHFFFAOYSA-N
10481
C(=N)(N)N.[N+](=O)(O)[O-]
CH6N4O3
MFCD00013028
guanidine nitrate, guanidinium nitrate, guanidine; nitric acid, guanidine, mononitrate, guanidine, nitrate, guanidine mononitrate, guanidine nitrate 1:1, guanidinenitrate, pubchem21372, acmc-209kpa
guanidine;nitric acid

Specifications

506-93-4
1.44
Odorless
MFCD00013028
UN1467
14,4562
CNUNWZZSUJPAHX-UHFFFAOYSA-N
guanidine;nitric acid
10481
98%
213°C to 216°C
4.9
CH6N4O3
250 g
3596600
guanidine nitrate, guanidinium nitrate, guanidine; nitric acid, guanidine, mononitrate, guanidine, nitrate, guanidine mononitrate, guanidine nitrate 1:1, guanidinenitrate, pubchem21372, acmc-209kpa
C(=N)(N)N.[N+](=O)(O)[O-]
122.084
122.08
Guanidine nitrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H302-H315-H319
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P210-P220-P221-P261-P264b-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P370+P380+P375-P372-P380-P501c

H272-H302-H315-H319-H350-X

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: GUANIDINE NITRATE

EINECSNumber : 208-060-1

RTECSNumber : MF4350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only