missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Guanosine 5'-monophosphate, disodium salt hydrate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 226600250

 View more versions of this product

Catalog No. AC226600250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Guanosine 5′-monophosphate,disodium salt hydrate
>300°C
25% max. (120°C, 4 hrs)
97%
C10H12N5Na2O8P
25 g
14, 4570
Solubility in water: >50g/l water
PVBRXXAAPNGWGE-KHRSEZDTNA-L
disodium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
21712
97%
5550-12-9
White
Authentic
Glass bottle
MFCD00151238
10, 279
methoxyphenamine hydrochloride, methoxyphenamine hcl, orthoxine hydrochloride, ortodrinex hydrochloride, methoxyphenaminium chloride, methoxiphenadrin hydrochloride, o-methoxymethamphetamine hydrochloride, 1-2-methoxyphenyl propan-2-yl methyl amine hydrochloride, 1-2-methoxyphenyl-n-methylpropan-2-amine hydrochloride
97% min. (lambda max. 252nm,pH 7) molar absorptivity 13700+/-4%,pH 7
[Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1
407.19
407.19
Crystalline Powder

Chemical Identifiers

5550-12-9
407.19
PVBRXXAAPNGWGE-KHRSEZDTNA-L
disodium;[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
C10H12N5Na2O8P
MFCD00151238
21712
[Na+].[Na+].NC1=NC2=C(N=CN2[C@@H]2O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]2O)C(=O)N1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-914-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only