missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
GVS Polyvinylidene Fluoride Hybridization and Transfer Membranes
Designed for Western transfers, amino acid analyses, and protein sequencing procedures
$259.81 - $1130.16
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Format | Pore Size | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Width (Metric) | Format | Pore Size | Price | Quantity | ||
PV4HY320F5
|
GVS
1212637 |
20 cm | 20 cm | Sheet | 0.45 μm |
Pack of 5 Each for $374.22
|
|
||
PV4HY315F5
|
GVS
1212636 |
15 cm | 15 cm | Sheet | 0.45 μm |
Pack of 5 Each for $259.81
|
|
||
PV4HY00010
|
GVS
1212639 |
30 cm | 3 m | Roll | 0.45 μm |
Each for $1,130.16
|
|
||
PV4HY910F0
|
GVS
1214558 |
9 cm | 10 cm | Sheet | 0.45 μm |
Pack of 10 Each for $323.06
|
|
||
PV4HY310F0
|
GVS
1212644 |
10 cm | 10 cm | Sheet | 0.45 μm |
Pack of 10 Each for $331.88
|
|
||
Description
- Compatible with most commonly used stains
- Won't degrade, distort or shrink in presence of high concentrations of methanol
- Efficient protein transfer from 5 to 700kDa
- Detection limit, 0.005μg/cm2
- Packaged in a wide variety of precut circles and sheets to fit Petri dishes and transfer/hybridization apparatus
- Also available in rolls for cutting custom-sized membranes
Manufactured by GVS Maine.
Specifications
PVDF-Plus Polyvinylidene Fluoride |