GVS Polyvinylidene Fluoride Hybridization and Transfer Membranes
Designed for Western transfers, amino acid analyses, and protein sequencing procedures
Supplier: GVS 1212644
Description
- Compatible with most commonly used stains
- Won't degrade, distort or shrink in presence of high concentrations of methanol
- Efficient protein transfer from 5 to 700kDa
- Detection limit, 0.005μg/cm2
- Packaged in a wide variety of precut circles and sheets to fit Petri dishes and transfer/hybridization apparatus
- Also available in rolls for cutting custom-sized membranes
Manufactured by GVS Maine.
Specifications
10 cm | |
Sheet | |
0.45 μm |
10 cm | |
PVDF-Plus Polyvinylidene Fluoride |