missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

H-B Instrument™ FRIO-Temp™ Precision Liquid-in-Glass Verification Thermometers

To verify sample temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST™ and DAkkS.

 View more versions of this product

Catalog No. 13-202-510


Explore available promotions

Description

Description

To verify sample temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST™ and DAkkS.

Each item features a fluoropolymer coating for safety, individual serial numbers and a multilanguage (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy indicating accuracy traceable to NIST and DAkkS.

Tested and calibrated in triple-accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment who calibration is traceable to NIST and DAkkS.

Certifications

NIST, DAkkS

Warranty

Five-year limited. See hbinstrument.com/conditions.php for details.

Specifications

Specifications

ISO/IEC 17025:2005, A2LA, NIST, DAkkS/PTB
Precision
135 mm
Enviro-Safe™ Liquid
1°C
-30°C to +1°C
0.5°C
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
5.3 in.
Glass
Verification Thermometer
-30°C
Freezer
0.5°
Videos
SDS
Documents

Documents

Promotions

Promotions