Learn More
H-B Instrument™ FRIO-Temp™ Precision Liquid-in-Glass Verification Thermometers
To verify sample temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST™ and DAkkS.
$118.50 - $287.86
Catalog Number | Mfr. No. | For Use With (Equipment) | Length (Metric) Overall | Accuracy | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Length (Metric) Overall | Accuracy | Price | Quantity | |||
13-201-469
|
H-B Instrument™
HBB61004110 |
Incubator | 135 mm | 1°F |
Each for $241.69
|
|
|||
13201552
|
H-B Instrument™
B610040500RMD |
Incubator | 135 mm | 0.5°C |
Each for $118.50
|
|
|||
13201551
|
H-B Instrument™
B610040400RMD |
Blood Bank | 135 mm | 0.5°C |
Each for $131.84
|
|
|||
13-202-508
|
N/A | Refrigerator | 125 mm | 0.5°C |
Each for $146.14
|
|
|||
13-202-510
|
N/A | Freezer | 135 mm | 0.5°C |
Each for $146.14
|
|
|||
13-202-507
|
N/A | Ultra Low Freezer | 145 mm | ±1°C, 2°C from -1/-40°C, and 2.5°C below -40°C |
Each for $287.86
|
|
|||
Description
To verify sample temperatures in freezers, refrigerators, incubators and ovens with traceability to NIST™ and DAkkS.
Each item features a fluoropolymer coating for safety, individual serial numbers and a multilanguage (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy indicating accuracy traceable to NIST and DAkkS.
Tested and calibrated in triple-accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment who calibration is traceable to NIST and DAkkS.
Certifications
NIST, DAkkS
Warranty
Five-year limited. See hbinstrument.com/conditions.php for details.