Learn More
Hafnium(IV) isopropoxide isopropanol adduct, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5228822
Description
Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.
Solubility
It reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents.
Specifications
Hafnium(IV) isopropoxide isopropanol adduct | |
C12H32HfO4 | |
MFCD00070460 | |
UN3181 | |
propan-2-ol-hafnium 4/1, 2-propanol, hafnium 4+ salt 9ci | |
HMKGKDSPHSNMTM-UHFFFAOYSA-N | |
hafnium;propan-2-ol | |
74764872 | |
99% |
2171-99-5 | |
Hf(OOCH(CH3)2)4·HOCH(CH3)2 | |
100 g | |
Moisture sensitive | |
It reacts with water. | |
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Hf] | |
418.874 | |
474.94 |
Chemical Identifiers
2171-99-5 | |
418.874 | |
HMKGKDSPHSNMTM-UHFFFAOYSA-N | |
74764872 | |
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Hf] |
C12H32HfO4 | |
MFCD00070460 | |
propan-2-ol-hafnium 4/1, 2-propanol, hafnium 4+ salt 9ci | |
hafnium;propan-2-ol |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only