missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hafnium(IV) isopropoxide isopropanol adduct, 99%, Thermo Scientific Chemicals

$2206.63 - $2206.63

Chemical Identifiers

CAS 2171-99-5
Molecular Formula C12H32HfO4
Molecular Weight (g/mol) 418.874
MDL Number MFCD00070460
InChI Key HMKGKDSPHSNMTM-UHFFFAOYSA-N
Synonym propan-2-ol-hafnium 4/1, 2-propanol, hafnium 4+ salt 9ci
PubChem CID 74764872
IUPAC Name hafnium;propan-2-ol
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Hf]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5228822
View Documents
Thermo Scientific Chemicals
H5228822
100 g
Each for $2,206.63
 
Description

Description

Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents.
Specifications

Chemical Identifiers

2171-99-5
418.874
HMKGKDSPHSNMTM-UHFFFAOYSA-N
74764872
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Hf]
C12H32HfO4
MFCD00070460
propan-2-ol-hafnium 4/1, 2-propanol, hafnium 4+ salt 9ci
hafnium;propan-2-ol

Specifications

2171-99-5
Hf(OOCH(CH3)2)4·HOCH(CH3)2
100 g
Moisture sensitive
It reacts with water.
CC(C)O.CC(C)O.CC(C)O.CC(C)O.[Hf]
418.874
474.94
Hafnium(IV) isopropoxide isopropanol adduct
C12H32HfO4
MFCD00070460
UN3181
propan-2-ol-hafnium 4/1, 2-propanol, hafnium 4+ salt 9ci
HMKGKDSPHSNMTM-UHFFFAOYSA-N
hafnium;propan-2-ol
74764872
99%
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only