missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Hafnium Standard For ICP, 1000μg/mL in 2% HCl, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$286.00 - $533.56

Identifiants chimiques

CAS 7440-58-6
Molecular Formula Hf
Molecular Weight (g/mol) 178.49
MDL Number MFCD00011032
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
PubChem CID 23986
ChEBI CHEBI:33343
IUPAC Name hafnium
SMILES [Hf]
voir les spécifications

Comprend: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
PLHF1-2Y
Afficher les documents
SPEX CertiPrep Group LLC
PLHF12Y
125 mL Triple-leached LDPE bottle
chaque for $286.00
 
PLHF1-2X
Afficher les documents
SPEX CertiPrep Group LLC
PLHF12X
500 mL Triple-leached LDPE bottle
chaque for $533.56
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Hf1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Spécifications

Identifiants chimiques

7440-58-6
178.49
VBJZVLUMGGDVMO-UHFFFAOYSA-N
CHEBI:33343
[Hf]
Hf
MFCD00011032
23986
hafnium

Spécifications

7440-58-6,7647-01-0,7732-18-5
125 mL
0.1,2.0,97.9
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
MFCD00011032
[Hf]
178.49
CHEBI:33343
Matrix Concentration Cat. No.
2-5% HCl 1000μg/mL PLHF1-2Y, -2X 10, 000 PLHF1-3Y
AA, ICP
Triple-leached LDPE bottle
0.1,2.0,97.9
Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Certified Reference Material
Hf
VBJZVLUMGGDVMO-UHFFFAOYSA-N
hafnium
23986
Assurance
2% HCl
Hafnium Standard
Videos
Sécurité et manipulation

Sécurité et manipulation

Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

FDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only