Learn More
Hafnium Standard For ICP, 1000μg/mL in 2% HCl, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLHF12Y
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Hf | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Hafnium Standard | |
Triple-leached LDPE bottle | |
0.1,2.0,97.9 | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00011032 | |
[Hf] | |
178.49 | |
CHEBI:33343 | |
Matrix Concentration Cat. No. 2-5% HCl 1000μg/mL PLHF1-2Y, -2X 10, 000 PLHF1-3Y |
7440-58-6,7647-01-0,7732-18-5 | |
125 mL | |
0.1,2.0,97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Hf | |
VBJZVLUMGGDVMO-UHFFFAOYSA-N | |
hafnium | |
23986 | |
Assurance | |
2% HCl |
Chemical Identifiers
7440-58-6 | |
178.49 | |
VBJZVLUMGGDVMO-UHFFFAOYSA-N | |
CHEBI:33343 | |
[Hf] |
Hf | |
MFCD00011032 | |
23986 | |
hafnium |
Safety and Handling
Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only