Hamilton™ 1000 Series Gastight™ Syringes: Luer Lock Syringes, TLL Termination
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
$115.02 - $874.87
Specifications
For Use With (Application) Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments
Product Type Gastight Syringe
Needle Point Number No Needle Included
Quantity Ea.
View More Specs
Products 12
Catalog Number Mfr. No. Volume (Metric) Model Connection Type Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Model Connection Type Price Quantity  
14-815-193
View Documents
Hamilton™
HAM82527
25 mL 1025 Luer Lock with Slots
Each for $533.11
13-684-116A
View Documents
Hamilton™
HAM86020
100 mL 1100 Luer Lock
Each for $874.87
13-684-94
View Documents
Hamilton™
HAM81320
1 mL 1001 Luer Lock
Each for $115.02
13-684-95
View Documents
Hamilton™
HAM81420
2.5 mL 1002 Luer Lock
Each for $120.76
13-684-96
View Documents
Hamilton™
HAM81520
5 mL 1005 Luer Lock
Each for $142.32
13-684-97
View Documents
Hamilton™
HAM81620
10 mL 1010 Luer Lock
Each for $208.57
14-815-129
View Documents
Hamilton™
HAM81327
1 mL 1001 Luer Lock with Slots
Each for $141.94
14-815-154
View Documents
Hamilton™
HAM81427
2.5 mL 1002 Luer Lock with Slots
Each for $141.26
14-815-170
View Documents
Hamilton™
HAM81527
5 mL 1005 Luer Lock with Slots
Each for $176.49
14-815-184
View Documents
Hamilton™
HAM81627
10 mL 1010 Luer Lock with Slots
Each for $207.58
14-815-194
View Documents
Hamilton™
HAM82520
25 mL 1025 Luer Lock
Each for $562.43
14-815-195
View Documents
Hamilton™
HAM85020
50 mL 1050 Luer Lock
Each for $747.06
Description
Description
Mid-volume GASTIGHT syringe ideal for dispensing volumes from 100 μL up to 100 mL.
  • PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors
  • The TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations
  • The version with slots is used during the attachment to some instruments but does not effect the connection to needles
  • Autoclavable when disassembled, except on 25mL and greater syringes; repeated autoclaving will shorten syringe life
  • Needle point styles 4 and AS are available on request)
Specifications
Specifications
Ideal for manual and automated HPLC injections, commonly used in syringe pumps and other liquid handling instruments
Gastight Syringe
No Needle Included
Ea.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'