Hamilton™ 700 Microliter™ Syringes: LT Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
$68.09 - $197.13
Specifications
Autoclavable Autoclavable
Material PTFE (Tip), Stainless Steel (Plunger)
For Use With (Application) For dispensing low volumes to achieve the highest level of accuracy and precision possible
Product Type Microliter Syringe
Max. Temperature (English) 239°F
View More Specs
Products 6
Catalog Number Mfr. No. Volume (Metric) Diameter (English) Inner Diameter (Metric) Outer Max. Pressure Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Diameter (English) Inner Diameter (Metric) Outer Max. Pressure Price Quantity  
14-813-1
View Documents
Hamilton™
HAM80301
10 μL 0.019 in. 6.6 mm 2000 psi
Each for $68.09
14-813-18
View Documents
Hamilton™
HAM80401
25 μL 0.029 in. 7.75 mm 2000 psi
Each for $93.85
14-813-65
View Documents
Hamilton™
HAM80701
250 μL 0.091 in. 7.75 mm 1000 psi
Each for $169.30
14-813-81
View Documents
Hamilton™
HAM80801
500 μL 0.135 in. 7.75 mm 1000 psi
Each for $197.13
14-824-10
View Documents
Hamilton™
HAM80501
50 μL 0.041 in. 7.75 mm 2000 psi
Each for $119.60
14-824-11
View Documents
Hamilton™
HAM80601
100 μL 0.061 in. 7.75 mm 2000 psi
Each for $141.70
Description
Description
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
Specifications
Autoclavable
For dispensing low volumes to achieve the highest level of accuracy and precision possible
239°F
50°F
Ea.
PTFE (Tip), Stainless Steel (Plunger)
Microliter Syringe
115°C
No Needle Included
Luer
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'