Hamilton™ Luer Lock Adapters for Hamilton™Valves
Adapters are designed to work with Hamilton valves
Supplier:  Hamilton™ HAM35030
 View more versions of this product
Catalog No. 14-819-278
Explore available promotions
Description
Description
Fittings have a 1/4-28 thread connection on one side and either a male luer, female luer, or male luer lock on the other side.
Specifications
Specifications
Adapter with Male Luer Lock
1/4-28 Thread .059 in. (1.5mm)