Hamilton™ Luer Lock Adapters for Hamilton™Valves
Adapters are designed to work with Hamilton valves
$33.17 - $33.17
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
14-819-278
|
Hamilton™
HAM35030 |
Adapter with Male Luer Lock |
Each for $33.17
|
|
|||||
14-819-285
|
Hamilton™
HAM35031 |
Adapter with Female Luer Lock |
Each for $33.17
|
|
|||||
Description
Fittings have a 1/4-28 thread connection on one side and either a male luer, female luer, or male luer lock on the other side.
Specifications
1/4-28 Thread .059 in. (1.5mm) |