Hamilton™ Microlab™ 500 Diluter/Dispenser System Accessories
Tubing Assembly, PTFE, FEP
Supplier:  Hamilton™ HAM240130
 View more versions of this product
Catalog No. 14-819-158
Explore available promotions
Description
Description
For use with Hamilton Microlab 500 Diluter/Dispenser
Specifications
Specifications
Tubing Assembly
35.43 in.
Microlab 500 Diluter/Dispenser
18 G
FEP, PTFE
90 cm
Use with syringe volumes ≤1mL
702 μL