missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hamilton™ Microlab™ 500 Diluter/Dispenser System Accessories
Tubing Assembly, PTFE, FEP
$123.03 - $123.03
Specifications
Product Type | Tubing Assembly |
---|---|
Material | FEP, PTFE |
Overall Length (English) | 35.43 in. |
Overall Length (Metric) | 90 cm |
For Use With | Microlab 500 Diluter/Dispenser |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
14-819-158
|
Hamilton™
HAM240130 |
Tubing Assembly |
N/A
|
|
|||||
Description
For use with Hamilton Microlab 500 Diluter/Dispenser
Specifications
Tubing Assembly | |
35.43 in. | |
Microlab 500 Diluter/Dispenser |
FEP, PTFE | |
90 cm |