Handheld Electronic Leak Detector Accessories from Restek™
Protects data and analytical columns from costly repairs
Supplier: Chromatographic RK22658
Alert:
Electronic leak detector determines trace amounts of gas in a GC environment only. Unit is not designed for determining combustible gas leaks. Avoid using liquid leak detectors in a capillary system: liquids can be drawn into the system.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Includes: Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs)
Description
- Optimized sample flow path
- Sleek ergonomic design
- Handy probe storage
- Long-lasting battery: up to 6 hours of continuous use
- Automatic shut-off capabilities
- Soft-sided carrying case designed for storage in a smaller space
- Small probe detector useful to verify hard-to-reach leaks
Certifications
CE marked
Compliance
WEEE, RoHS
Specifications
Handheld Leak Detector | |
CE, WEEE, RoHS | |
Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs) |
Small Probe Adapter | |
Protects data and analytical columns from costly repairs |