Handheld Electronic Leak Detector Accessories from Restek™
Protects data and analytical columns from costly repairs
$98.98 - $217.72
Specifications
For Use With (Equipment) | Handheld Leak Detector |
---|---|
Certifications/Compliance | CE, WEEE, RoHS |
For Use With (Application) | Protects data and analytical columns from costly repairs |
Includes | Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs) |
Alert:
Electronic leak detector determines trace amounts of gas in a GC environment only. Unit is not designed for determining combustible gas leaks. Avoid using liquid leak detectors in a capillary system: liquids can be drawn into the system.
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
06-722-105
![]() |
Chromatographic
RK22657 |
Soft Side Carrying Case |
Each for $98.98
|
|
|||||
06-722-106
![]() |
Chromatographic
RK22658 |
Small Probe Adapter |
Each for $217.72
|
|
|||||
Description
- Optimized sample flow path
- Sleek ergonomic design
- Handy probe storage
- Long-lasting battery: up to 6 hours of continuous use
- Automatic shut-off capabilities
- Soft-sided carrying case designed for storage in a smaller space
- Small probe detector useful to verify hard-to-reach leaks
Certifications
CE marked
Compliance
WEEE, RoHS
Specifications
Handheld Leak Detector | |
CE, WEEE, RoHS | |
Protects data and analytical columns from costly repairs | |
Convenient hard-sided carrying case; Universal charger set (US, UK and European plugs) |