missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oakton™ Handheld pH Meters
High +/-0.01 pH accuracy at an economical price.
Supplier: Oakton™ 3561324
Includes: Meter includes protective rubber boot and batteries.
Kit includes meter, "all-in-one" single-junction pH electrode, pH/ATC probe, pH buffer solutions, sample bottles, rinse bottle, protective rubber boot, batteries and hard carrying case.
Description
- Simple push-button operation
- Automatic temperature compensation (ATC)
- Hold and Auto-off functions
- Built-in stand for benchtop or long-term use
- Protective boot helps shield meter from drops and dings
- Meter kits contain everything needed for calibration and measurement and is packaged in a hard carrying case
- Toggle between pH and temperature in °C with a press of a button
- Also features measurements in pH, mV, and °C for ORP (Redox) measurements
- Three-point pH calibration: standard US, NIST, and pure water; gives you high ±0.01 pH accuracy
- Calibration instructions printed on back of meter
- Automatically identifies the correct pH buffer for rapid calibration
Certifications
ISO 9001:2000, CE marked (Meters)
Warranty
Three years warranty (meter only)
Specifications
Meter, “all-in-one” single-junction pH electrode, pH/ATC probe, pH buffer solutions, sample bottles, rinse bottle, protective rubber boot, batteries and hard carrying case. | |
±1000 | |
1 to 3 | |
0.4 kg | |
Automatic from 0° to 100°C | |
0°C to 50°C | |
pH 6+ Meter Kit | |
14 cm | |
1.37 in. |
Simple push-button operation, automatic temperature compensation, hold and auto-off functions, built-in stand for benchtop or long-term use | |
1mV | |
USA, NIST, ISO 9001 2000, CE | |
0.9 lb. | |
32.0°F to 122.0°F | |
4 x AAA | |
>70 hr. | |
2.75 in. | |
3.5 cm |