missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oakton™ Handheld pH Meters
High +/-0.01 pH accuracy at an economical price.
$1,377.41 - $1,377.41
Specifications
Battery Life | >70 hr. |
---|---|
Battery Type | 4 x AAA |
For Use With (Application) | Simple push-button operation, automatic temperature compensation, hold and auto-off functions, built-in stand for benchtop or long-term use |
Height (English) | 1.37 in. |
Height (Metric) | 3.5 cm |
Catalog Number | Mfr. No. | Includes | Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Type | Price | Quantity | ||||
13-300-103
|
Oakton™
3561324 |
Meter, “all-in-one” single-junction pH electrode, pH/ATC probe, pH buffer solutions, sample bottles, rinse bottle, protective rubber boot, batteries and hard carrying case. | pH 6+ Meter Kit |
N/A
|
|
||||
Description
- Simple push-button operation
- Automatic temperature compensation (ATC)
- Hold and Auto-off functions
- Built-in stand for benchtop or long-term use
- Protective boot helps shield meter from drops and dings
- Meter kits contain everything needed for calibration and measurement and is packaged in a hard carrying case
- Toggle between pH and temperature in °C with a press of a button
- Also features measurements in pH, mV, and °C for ORP (Redox) measurements
- Three-point pH calibration: standard US, NIST, and pure water; gives you high ±0.01 pH accuracy
- Calibration instructions printed on back of meter
- Automatically identifies the correct pH buffer for rapid calibration
Certifications
ISO 9001:2000, CE marked (Meters)
Warranty
Three years warranty (meter only)
Specifications
>70 hr. | |
Simple push-button operation, automatic temperature compensation, hold and auto-off functions, built-in stand for benchtop or long-term use | |
3.5 cm | |
1mV | |
32.0°F to 122.0°F | |
0.9 lb. | |
2.75 in. |
4 x AAA | |
1.37 in. | |
14 cm | |
Automatic from 0° to 100°C | |
0°C to 50°C | |
0.4 kg |