Lide Labs Hansel™ Stains DFS Item
For the examination of the nasal secretions to differentiate an allergic condition from a bacterial infection
Supplier:  Lide Labs LIDEHS500
 View more versions of this product
Catalog No. 23233801
Explore available promotions
Description
Description
  • One-minute stain
  • For secretion cytology
  • Stain eosinophils red; neutrophils, epithelial (exfoliated cells) and bacteria blue; and mast cells or basophiles red and blue
  • Can be adapted for staining various secretions: urinary, conjunctival, salivary, gastrointestinal, nasal, sinus, aural and bronchial
  • Determine elevated levels of eosinophils in urine (eosinophiluria) which can aid in diagnosis of many associated clinical conditions
  • Cannot be used for blood staining
Specifications
Specifications
Hansel Stain
Secretion cytology stains eosinophils red, neutrophils, epithelial (exfoliated cells) and bacteria blue, and mast cells or basophiles red and blue
500mL