Lide Labs Hansel™ Stains DFS Item
For the examination of the nasal secretions to differentiate an allergic condition from a bacterial infection
$305.26 - $305.26
Specifications
Type Hansel Stain
For Use With (Application) Secretion cytology stains eosinophils red, neutrophils, epithelial (exfoliated cells) and bacteria blue, and mast cells or basophiles red and blue
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
23233801 DFS Item
View Documents
Lide Labs
LIDEHS500
500mL
Each for $305.26
Description
Description
  • One-minute stain
  • For secretion cytology
  • Stain eosinophils red; neutrophils, epithelial (exfoliated cells) and bacteria blue; and mast cells or basophiles red and blue
  • Can be adapted for staining various secretions: urinary, conjunctival, salivary, gastrointestinal, nasal, sinus, aural and bronchial
  • Determine elevated levels of eosinophils in urine (eosinophiluria) which can aid in diagnosis of many associated clinical conditions
  • Cannot be used for blood staining
Specifications
Specifications
Hansel Stain
Secretion cytology stains eosinophils red, neutrophils, epithelial (exfoliated cells) and bacteria blue, and mast cells or basophiles red and blue
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'