missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Hard Hat Labels - Pressure Sensitive Vinyl
Pressure Sensitive Vinyl (B-946) is an indoor/outdoor marking film ideal for pipe marking, arrow tape, vehicle placards, pedestal markers, warning panels and safety signs.
$5.76 - $6.89
Specifications
Autoclavable | Not Autoclavable |
---|---|
Description | Label |
Material | Vinyl |
Languages | English |
No. per Pack | 4 Labels/Card |
Catalog Number | Mfr. No. | Display Text | Color | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Display Text | Color | Price | Quantity | ||||
19-110-268
|
Brady™
42231 |
BE CAREFUL | White on Green |
Each for $6.89
|
|
||||
19110265
|
Brady™
42228 |
AUTHORIZED OPERATOR FORK LIFT | Black on Yellow |
Each for $6.89
|
|
||||
19-110-266
|
Brady™
42229 |
AUTHORIZED FORKLIFT DRIVER | Black on Yellow |
Each for $6.89
|
|
||||
19-110-270
|
Brady™
42233 |
CERTIFIED FORKLIFT DRIVER | Black on Yellow |
Each for $6.89
|
|
||||
19-110-271
|
Brady™
42234 |
CPR FIRST AID CERTIFIED AND TRAINED | White on Red |
Each for $6.89
|
|
||||
19-110-272
|
Brady™
42238 |
EMERGENCY RESPONSE TEAM | Black/Red on White |
Each for $6.89
|
|
||||
19-110-274
|
Brady™
42242 |
FIRST... SAFETY ...LAST | Black/Red on White |
Each for $6.89
|
|
||||
19-110-276
|
Brady™
42244 |
HEALTH & SAFETY COMMITTEE | Green on White |
Each for $6.89
|
|
||||
19-110-275
|
Brady™
42243 |
FORKLIFT SAFETY TRAINED | Black on Yellow |
Each for $6.89
|
|
||||
19-110-277
|
Brady™
42245 |
HEALTH & SAFETY COMMITTEE MEMBER | Green on White |
Each for $6.89
|
|
||||
19-110-278
|
Brady™
42247 |
KNOCK OUT... ACCIDENTS | Black/Brown on White |
Each for $6.89
|
|
||||
19-110-279
|
Brady™
42248 |
LOCK-OUT TAG-OUT TRAINED | Black/Red on White |
Each for $6.89
|
|
||||
19-110-280
|
Brady™
42257 |
OUR AIM... NO ACCIDENTS | Black/Red on White |
Each for $6.89
|
|
||||
19-110-281
|
Brady™
42261 |
QUALITY STARTS WITH ME! | Red on White |
Each for $6.89
|
|
||||
19-110-282
|
Brady™
42262 |
QUALIFIED CRANE OPERATOR | Black/Orange on White |
Each for $6.89
|
|
||||
19-110-285
|
Brady™
42271 |
SAFETY... IT'S MY JOB | Black/Red on White |
Each for $6.89
|
|
||||
19-110-283
|
Brady™
42263 |
QUALIFIED FORK LIFT OPERATOR | Black/Yellow on White |
Each for $6.89
|
|
||||
19-110-284
|
Brady™
42269 |
SAFETY BEGINS WITH TEAMWORK | Black/Yellow on White |
Each for $6.89
|
|
||||
19-110-286
|
Brady™
42272 |
SAFETY IS NOT AN ACCIDENT | Black/Red on Yellow |
Each for $6.89
|
|
||||
19-110-287
|
Brady™
42273 |
SAFETY NEVER HURTS | Black/Red on Yellow |
Each for $6.89
|
|
||||
19-110-288
|
Brady™
42283 |
THINK SAFETY 1st | Red on White |
Each for $6.89
|
|
||||
19-110-289
|
Brady™
42284 |
VISITOR | Black on White |
Each for $6.89
|
|
||||
19-110-290
|
Brady™
42290 |
YOUR FUTURE BRIGHTENS WITH SAFETY | Black/Yellow on White |
Each for $6.89
|
|
||||
19-110-291
|
Brady™
42293 |
CERTIFIED CRANE OPERATOR | Black/Red on White |
Each for $6.89
|
|
||||
19-110-292
|
Brady™
BRD42294 |
CPR TRAINED CERTIFIED | Black/Blue/Red on White |
Each for $6.89
|
|
||||
19-110-293
|
Brady™
42541 |
RIGHT TO KNOW TRAINED | Black/Blue/Red/Yellow on White |
Each for $6.89
|
|
||||
19110295
|
Brady™
BRD49550 |
ACCIDENT FREE AWARD 1 YEAR AWARD | Green on White |
Each for $5.76
|
|
||||
19-110-296
|
Brady™
49551 |
ACCIDENT FREE 5 YEARS AWARD | Green on White |
Each for $5.76
|
|
||||
19-110-297
|
Brady™
49552 |
SAFE WORKER NO ACCIDENTS/1 YEAR | Green on White |
Each for $5.76
|
|
||||
19-110-298
|
Brady™
49553 |
SAFE WORKER NO ACCIDENTS 5 YEARS | Green on White |
Each for $5.76
|
|
||||
19110299
|
Brady™
BRD49555 |
SAFETY AWARD WINNER | Green on White |
Each for $5.76
|
|
||||
19110300
|
Brady™
49556 |
EMERGENCY RESPONSE TEAM | Red on White |
Each for $5.76
|
|
||||
19-110-301
|
Brady™
49557 |
SAFE DRIVER | Green on White |
Each for $5.76
|
|
||||
19-110-303
|
Brady™
49559 |
MEDICAL EMERGENCY TEAM | Red on White |
Each for $5.76
|
|
||||
19-110-302
|
Brady™
49558 |
HAZARDOUS MATERIALS MANAGEMENT | Red on White |
Each for $5.76
|
|
||||
19-110-304
|
Brady™
49560 |
CERTIFIED CRANE OPERATOR | Black on White |
Each for $5.76
|
|
||||
19-110-305
|
Brady™
49561 |
CPR CERTIFIED | Red on White |
Each for $5.76
|
|
||||
19-110-306
|
Brady™
49562 |
CHEMICAL EMERGENCY TEAM | Red on White |
Each for $5.76
|
|
||||
19-110-308
|
Brady™
49564 |
SAFETY SUPERVISOR | Green on White |
Each for $5.76
|
|
||||
19-110-307
|
Brady™
49563 |
GOOD HOUSEKEEPING COMMITTEE | Blue on White |
Each for $5.76
|
|
||||
19-110-310
|
Brady™
49566 |
RESCUE TEAM | Red on White |
Each for $5.76
|
|
||||
19-110-309
|
Brady™
49565 |
CERTIFIED FIRST AID | Red on White |
Each for $5.76
|
|
||||
19-110-314
|
Brady™
49570 |
EMERGENCY MEDICAL TECHNICIAN | Black on White |
Each for $5.76
|
|
||||
19-110-313
|
Brady™
49569 |
HEALTH AND SAFETY/COMMITTEE MEMBER | Green on White |
Each for $5.76
|
|
||||
19-110-315
|
Brady™
49571 |
PLANT SAFETY MEMBER | Green on White |
Each for $5.76
|
|
||||
19-110-316
|
Brady™
49572 |
SAFETY OBSERVER | Green on White |
Each for $5.76
|
|
||||
19-110-317
|
Brady™
49573 |
SAFETY REPRESENTATIVE | Green on White |
Each for $5.76
|
|
||||
19-110-318
|
Brady™
49574 |
ACCIDENT PREVENTION MEMBER | Red on White |
Each for $5.76
|
|
||||
19-110-319
|
Brady™
49576 |
BASIC LIFE SUPPORT CERTIFIED CPR | Red on White |
Each for $5.76
|
|
||||
19-110-320
|
Brady™
49577 |
SAFE WORKER NO ACCIDENTS | Green on White |
Each for $5.76
|
|
||||
19-110-321
|
Brady™
49578 |
HAZ MAT SQUAD | Red on White |
Each for $5.76
|
|
||||
19-110-322
|
Brady™
49579 |
ERGONOMICS COMMITTEE | Green on White |
Each for $5.76
|
|
||||
19-110-326
|
Brady™
76305 |
MEMBER FIRE BRIGADE | Red on White |
Each for $5.76
|
|
||||
19-110-327
|
Brady™
76306 |
FIRST AID SQUAD | Green on White |
Each for $5.76
|
|
||||
Description
- High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
- Good chemical, oil and water resistance
- Average outdoor durability 8-10 years
- Service temperature of -40°F to 180°F (-40°C to 82°C)
Specifications
Not Autoclavable | |
Vinyl | |
4 Labels/Card | |
2 in. |
Label | |
English | |
1 Card |