missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Hazardous Waste Label: HAZARDOUS WASTE ACCUMULATION START DATE CONTENTS...
Identify hazardous waste materials for transportation off-site. Brady™ Hazardous Waste Labels are made of vinyl and comply with all current federal regulations regarding identification of hazardous waste material transportation.
Supplier: Brady™ 121063
Description
- To identify containers, cylinders and drums holding hazardous waste
- Made of B-7569 vinyl
- Available in size (W x H): 15.2 x 10.16cm (6 x 4 in.)
Specifications
Acrylic, Permanent | |
Vinyl (B-7569) | |
HAZARDOUS WASTE ACCUMULATION START DATE CONTENTS...etc | |
Square | |
0.004 in. |
Black, Red, White | |
Hazardous Waste Label | |
100/Pk. | |
6 x 4 in. (W x H) |