missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Hazardous Waste Label: HAZARDOUS WASTE ACCUMULATION START DATE CONTENTS...
Identify hazardous waste materials for transportation off-site. Brady™ Hazardous Waste Labels are made of vinyl and comply with all current federal regulations regarding identification of hazardous waste material transportation.
$120.65 - $149.22
Specifications
Adhesive Type | Acrylic, Permanent |
---|---|
Color | Black, Red, White |
Material | Vinyl (B-7569) |
Product Type | Hazardous Waste Label |
Display Text | HAZARDOUS WASTE ACCUMULATION START DATE CONTENTS...etc |
Catalog Number | Mfr. No. | Size (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size (English) | Price | Quantity | |||||
19-110-962
|
Brady™
121063 |
6 x 4 in. (W x H) |
Pack of 100 Each for $120.65
|
|
|||||
19110963
|
Brady™
BRD121064 |
6 x 4 in. (W x H) |
Pack of 50 Each for $149.22
|
|
|||||
Description
- To identify containers, cylinders and drums holding hazardous waste
- Made of B-7569 vinyl
- Available in size (W x H): 15.2 x 10.16cm (6 x 4 in.)
Specifications
Acrylic, Permanent | |
Vinyl (B-7569) | |
HAZARDOUS WASTE ACCUMULATION START DATE CONTENTS...etc | |
6 x 4 in. (W x H) |
Black, Red, White | |
Hazardous Waste Label | |
Square | |
0.004 in. |