missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Hazardous Waste Label: NON-HAZARDOUS WASTE
Identify hazardous waste materials for transportation off-site. Brady™ Hazardous Waste Labels are made of vinyl and comply with all current federal regulations regarding identification of hazardous waste material transportation.
Supplier: Brady™ 121159
Description
- To identify containers, cylinders and drums holding hazardous waste
- Made of B-7569 vinyl
- Available in size (W x H): 15.2 x 15.2cm (6 x 6 in.)
Specifications
| Acrylic, Permanent | |
| Vinyl (B-7569) | |
| NON-REGULATED WASTE | |
| Square | |
| 0.004 in. |
| White, Green | |
| Hazardous Waste Label | |
| 50/Pk. | |
| 6 x 6 in. |