missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments

Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.

Supplier:  Thermo Scientific™ AB3720

 View more versions of this product

Catalog No. AB3720


Explore available promotions

Description

Description

Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.

Features:

  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications

Specifications

Foil
Aluminum
Not Autoclavable
1
78 mm x 610 m
General; Long term
ALPS5000 & ALPS 3000
Abgene
No
-80°C to +120°C
Polypropylene, Polyethylene
Yes
Life Science Reagents Resistant, DMSO Resistant
Non-sterile
96
1
Ideal for compound management and high throughput screening applications, suitable for PCR
Roll
EasyPierce 20 μm Thin Foil
Polypropylene
Yes
All well shapes
General
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions