Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments
Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.
$6907.58 - $9576.79
Specifications
Format Roll
View More Specs
Products 2
Catalog Number Mfr. No. Material Product Type Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Material Product Type Dimensions (L x W) Price Quantity  
AB-3559
View Documents
Thermo Scientific™
AB3559
Aluminum ThermoSeal 78 mm x 370 m
Each for $6,907.58
AB3720
View Documents
Thermo Scientific™
AB3720
Aluminum EasyPierce 20 μm Thin Foil 78 mm x 610 m
Each for $9,576.79
Description
Description
Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.
Features:
  • Quick operation
  • Safe for temperature-sensitive samples
  • Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications
Specifications
Roll
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'