Learn More
Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments
Save time and costs while improving sealing integrity with these microplate heat sealing instruments. Offers the capability of retrieving samples and then re-sealing the plate.
Supplier: Thermo Scientific™ AB3559
Description
Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.
Features:
- Quick operation
- Safe for temperature-sensitive samples
- Designed for reliable performance with Thermo Scientific ALPS instruments, including the, ALPS 3000, ALPS 25 and ALPS 50 V
Specifications
| Foil | |
| Aluminum | |
| Not Autoclavable | |
| 1 | |
| 78 mm x 370 m | |
| General; Long term storage including storage at low temperatures | |
| ALPS 500 & ALPS 3000 | |
| Abgene | |
| No | |
| -80°C to +120°C | |
| Polypropylene | |
| Very Strong Seal that requires stripper tool for peeling |
| Life Science Reagents Resistant, DMSO Resistant | |
| Non-sterile | |
| 96 | |
| 1 | |
| Transportation and high temperature applications such as PCR | |
| Roll | |
| ThermoSeal | |
| Polypropylene | |
| Yes | |
| All well shapes | |
| General |