Synthware™ Heavy Wall Pressure Vessels
Vessels feature an internal thread for use with Teflon™ bushing as a pressure seal. Unique design allows the vessel to be loosened and tightened by hand for easy access.
Supplier: Synthware™ P160702
Description
- Internal thread for use with Teflon™ bushing as a pressure seal
- Vessel can be loosened/tightened by hand for easy access
- O-ring is positioned on the bushing above the thread for improved sealing
- Complete apparatus includes vessel, Teflon™ bushing, and EP™ O-ring
- Ideal for pressure reactions and applications such as polymerization and halogen analysis by thermal decomposition in the presence of lime
Specifications
Vessel, Teflon™ Bushing, EP™ O-Ring | |
Pressure Reactions, Polymerization, and Halogen Analysis | |
#7 | |
45 mm |
Cylindrical | |
Borosilicate Glass | |
2 mL | |
13 mm |