missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hematein, Thermo Scientific Chemicals
$134.42 - $446.14
Chemical Identifiers
CAS | 475-25-2 |
---|---|
Molecular Formula | C16H12O6 |
Molecular Weight (g/mol) | 300.27 |
MDL Number | MFCD00036187 |
InChI Key | HLUCICHZHWJHLL-UHFFFAOYNA-N |
Synonym | hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein |
PubChem CID | 10138 |
IUPAC Name | 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one |
SMILES | OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC370840050
|
Thermo Scientific Chemicals
370840050 |
5 g | Glass bottle |
N/A
|
|
||||
AC370840250
|
Thermo Scientific Chemicals
370840250 |
25 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
475-25-2 | |
300.27 | |
HLUCICHZHWJHLL-UHFFFAOYNA-N | |
10138 | |
OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O |
C16H12O6 | |
MFCD00036187 | |
hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein | |
3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one |
Specifications
475-25-2 | |
Brown | |
Glass bottle | |
MFCD00036187 | |
18,IV,3343 | |
hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein | |
HLUCICHZHWJHLL-UHFFFAOYNA-N | |
3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one | |
10138 | |
Crystalline Powder |
180°C | |
Authentic | |
C16H12O6 | |
5 g | |
15,4669 | |
Solubility in water: very slightly soluble. Other solubilities: slightly solubel in ethanol,ether,soluble in alkaline solutions,insoluble in benzene,chloroform | |
OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O | |
300.27 | |
300.26 | |
Hematein |
Safety and Handling
EINECSNumber : 207-492-8
RUO – Research Use Only