missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hematoxylin Hydrate MP Biomedicals

Supplier:  MP Biomedicals Inc 0210192280

 View more versions of this product

Catalog No. ICN10192280


Explore available promotions

Description

Description

Specifications

Specifications

Hematoxylin
517-28-2
MFCD00078111
WZUVPPKBWHMQCE-WKTCHCBJNA-N
(1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
45029742
100 g
Hydrate
C16H14O6
Natural Black I
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
302.28
356.4

Chemical Identifiers

517-28-2
302.28
WZUVPPKBWHMQCE-WKTCHCBJNA-N
45029742
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
C16H14O6
MFCD00078111
Natural Black I
(1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
Videos
SDS
Documents

Documents

Promotions

Promotions