missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hematoxylin Hydrate MP Biomedicals

$596.24 - $596.24

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black I
PubChem CID 45029742
IUPAC Name (1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10192280
View Documents
MP Biomedicals Inc
0210192280
100 g
Each for $596.24
 
Description

Description

Specifications

Chemical Identifiers

517-28-2
302.28
WZUVPPKBWHMQCE-WKTCHCBJNA-N
45029742
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
C16H14O6
MFCD00078111
Natural Black I
(1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol

Specifications

517-28-2
MFCD00078111
WZUVPPKBWHMQCE-WKTCHCBJNA-N
(1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
45029742
100 g
C16H14O6
Natural Black I
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
302.28
356.4
Hematoxylin, Hydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'