missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HEPES sodium salt, >98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210559380

 View more versions of this product

Catalog No. ICN10559380


Explore available promotions

Description

Description

Specifications

Specifications

HEPES sodium salt
White
100 g
C8H17N2NaO4S
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
260.28
CHEBI:46758
75277-39-3
Powder
>98%
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
2724248
260.3

Chemical Identifiers

75277-39-3
260.28
RDZTWEVXRGYCFV-UHFFFAOYSA-M
2724248
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
C8H17N2NaO4S
MFCD00036463
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
CHEBI:46758
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 278-169-7

Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.

SDS
Documents

Documents

Promotions

Promotions