missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HEPES sodium salt, >98%, MP Biomedicals™

$90.71 - $1,258.78

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10559325
View Documents
MP Biomedicals Inc
0210559325
25 g N/A
 
ICN10559380
View Documents
MP Biomedicals Inc
0210559380
100 g N/A
 
ICN10559383
View Documents
MP Biomedicals Inc
0210559383
250 g N/A
 
ICN10559391
View Documents
MP Biomedicals Inc
0210559391
1 kg N/A
 
Description

Description

Specifications

Chemical Identifiers

75277-39-3
260.28
RDZTWEVXRGYCFV-UHFFFAOYSA-M
2724248
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
C8H17N2NaO4S
MFCD00036463
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
CHEBI:46758
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

75277-39-3
Powder
>98%
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
2724248
260.3
White
25 g
C8H17N2NaO4S
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
260.28
CHEBI:46758
HEPES sodium salt
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 278-169-7

Recommended Storage : Store at Room Temperature (15 to 30°C). Store Desiccated.

SDS
missing translation for 'documents'

missing translation for 'documents'