missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HEPES sodium salt, ≥99%, MP Biomedicals™
HEPES is widely used in many biochemical reactions and as a buffering agent in some cell culture media. It is used in a protein deposition technique in electron microscopy because it did not affect metal substrates
Supplier: MP Biomedicals Inc 0219482825
Specifications
HEPES sodium salt | |
White | |
25 g | |
C8H17N2NaO4S | |
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt | |
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 | |
260.28 | |
CHEBI:46758 |
75277-39-3 | |
Powder | |
≥99% | |
MFCD00036463 | |
RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate | |
2724248 | |
260.3 |
Chemical Identifiers
75277-39-3 | |
260.28 | |
RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
2724248 | |
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
C8H17N2NaO4S | |
MFCD00036463 | |
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt | |
CHEBI:46758 | |
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
Safety and Handling
EINECSNumber : 278-169-7