missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heptaldehyde, 95%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 120320010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Heptaldehyde | |
111-71-7 | |
0.8200g/mL | |
48°C | |
94% min. (GC) | |
C7H14O | |
CH3(CH2)5CHO | |
1 L | |
0.82 | |
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal | |
FXHGMKSSBGDXIY-UHFFFAOYSA-N | |
heptanal | |
100ppm hydroquinone | |
0.977 mPa.s (15°C) | |
114.19 | |
Powder, Crystals or Chunks |
95% | |
-42.0°C | |
150.0°C | |
Authentic | |
Glass bottle | |
1.4115 to 1.4135 | |
MFCD00007028 | |
01, II, 750 | |
15, 4693 | |
Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol | |
CCCCCCC=O | |
114.19 | |
8130 | |
CHEBI:34787 | |
95% |
Chemical Identifiers
111-71-7 | |
114.19 | |
FXHGMKSSBGDXIY-UHFFFAOYSA-N | |
8130 | |
heptanal |
C7H14O | |
MFCD00007028 | |
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal | |
CHEBI:34787 | |
CCCCCCC=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 203-898-4
RTECSNumber : MI6900000
TSCA : TSCA
RUO – Research Use Only