missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptaldehyde, 95%, stabilized, Thermo Scientific Chemicals

$41.26 - $183.82

Chemical Identifiers

CAS 111-71-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00007028
InChI Key FXHGMKSSBGDXIY-UHFFFAOYSA-N
Synonym heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
PubChem CID 8130
ChEBI CHEBI:34787
IUPAC Name heptanal
SMILES CCCCCCC=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120320500
SDS View Documents
Thermo Scientific Chemicals
120320500
50 mL Glass bottle
Each for $41.26
 
AC120322500
SDS View Documents
Thermo Scientific Chemicals
120322500
250 mL Glass bottle
Each for $61.20
 
AC120320010
SDS View Documents
Thermo Scientific Chemicals
120320010
1 L Glass bottle
Each for $183.82
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-71-7
114.19
FXHGMKSSBGDXIY-UHFFFAOYSA-N
8130
heptanal
C7H14O
MFCD00007028
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
CHEBI:34787
CCCCCCC=O

Specifications

111-71-7
Beige-Yellow to White
150.0°C
Authentic
Glass bottle
1.4115 to 1.4135
MFCD00007028
01, II, 750
15, 4693
Solubility in water: insoluble. Other solubilities: soluble in ether and alcohol
CCCCCCC=O
114.19
8130
CHEBI:34787
95%
Heptaldehyde, 95%
-42.0°C
0.8200g/mL
48°C
94% min. (GC)
C7H14O
CH3(CH2)5CHO
50 mL
0.82
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
FXHGMKSSBGDXIY-UHFFFAOYSA-N
heptanal
100ppm hydroquinone
0.977 mPa.s (15°C)
114.19
Powder, Crystals or Chunks
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

EINECSNumber : 203-898-4

RTECSNumber : MI6900000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only