missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heptane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%, Honeywell
Supplier: Honeywell Inc LP2064P
Specifications
Heptane | |
142-82-5 | |
0.684 g/cm3 | |
4 x 4 L | |
Poly Bottle | |
C7H16 | |
UN1206 | |
1730763 | |
0.03% in Water at 20°C | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
>99% |
ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0% | |
100.0000% | |
98.4°C | |
−4°C | |
by GC Analysis n-Heptane | |
MFCD00009544 | |
0.01% max. | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.21 | |
ACS Reagent |
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes