missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heptane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%, Honeywell
$851.70 - $7927.10
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6001038
|
Honeywell Inc
LP2064 |
4 x 4 L | Glass Bottle |
Case of 4 Each for $959.30
|
|
||||
6001039
|
Honeywell Inc
LP2064P |
4 x 4 L | Poly Bottle |
Case of 4 Each for $851.70
|
|
||||
6001036
|
Honeywell Inc
LP20619 |
19 L | Metal Can |
Each for $963.50
|
|
||||
6001037
|
Honeywell Chemicals
LP206204 |
204 L | Metal Drum |
Each for $7,927.10
|
|
||||
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
142-82-5 | |
0.684 g/cm3 | |
4 x 4 L | |
Glass Bottle | |
MFCD00009544 | |
0.01% max. | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.21 | |
ACS Reagent | |
by GC Analysis n-Heptane |
100.0000% | |
98.4°C | |
−4°C | |
C7H16 | |
UN1206 | |
1730763 | |
0.03% in Water at 20°C | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
>99% | |
Heptane, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0% |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Heptanes