missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexamethylenetetramine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 120610010

 View more versions of this product

Catalog No. AC120610010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexamethylenetetramine
260.0°C
1.3300g/mL
250°C
99%
C6H12N4
01,583
1.33
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C)
C1N2CN3CN1CN(C2)C3
4101
140.19
Crystals or Powder
100-97-0
White to Yellow
7 to 10 (1% aq. soln.)
Authentic
Plastic Bottle
1 kg
01,427; 02,208; 04,243; 09,234
15,6038
263.0 °C
VKYKSIONXSXAKP-UHFFFAOYSA-N
140.19
CHEBI:6824
99%

Chemical Identifiers

100-97-0
140.19
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
CHEBI:6824
C6H12N4
VKYKSIONXSXAKP-UHFFFAOYSA-N
4101
C1N2CN3CN1CN(C2)C3
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Flammable solid.

GHS P Statement
Wear protective gloves/protective clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

EINECSNumber : 202-905-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only