missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexamethylenetetramine, 99%, Thermo Scientific Chemicals

$86.41 - $615.33

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
Synonym methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
PubChem CID 4101
ChEBI CHEBI:6824
SMILES C1N2CN3CN1CN(C2)C3
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120610010
SDS View Documents
Thermo Scientific Chemicals
120610010
1 kg Plastic Bottle
Each for $86.41
 
AC120610025
SDS View Documents
Thermo Scientific Chemicals
120610025
2.5 kg Plastic Bottle
Each for $186.37
 
AC120610100
SDS View Documents
Thermo Scientific Chemicals
120610100
10 kg Plastic drum N/A
N/A
Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-97-0
140.19
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
CHEBI:6824
C6H12N4
VKYKSIONXSXAKP-UHFFFAOYSA-N
4101
C1N2CN3CN1CN(C2)C3

Specifications

100-97-0
White to Yellow
7 to 10 (1% aq. soln.)
Authentic
Plastic Bottle
1 kg
01,427; 02,208; 04,243; 09,234
15,6038
263.0 °C
VKYKSIONXSXAKP-UHFFFAOYSA-N
140.19
CHEBI:6824
99%
Hexamethylenetetramine
260.0°C
1.3300g/mL
250°C
99%
C6H12N4
01,583
1.33
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C)
C1N2CN3CN1CN(C2)C3
4101
140.19
Crystals or Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Flammable solid.

GHS P Statement
Wear protective gloves/protective clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Warning

EINECSNumber : 202-905-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only