missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexamethylenetetramine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 120610025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexamethylenetetramine | |
260.0°C | |
1.3300g/mL | |
250°C | |
99% | |
C6H12N4 | |
01,583 | |
1.33 | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C) | |
C1N2CN3CN1CN(C2)C3 | |
4101 | |
140.19 | |
Crystals or Powder |
100-97-0 | |
White to Yellow | |
7 to 10 (1% aq. soln.) | |
Authentic | |
Plastic Bottle | |
2.5 kg | |
01,427; 02,208; 04,243; 09,234 | |
15,6038 | |
263.0 °C | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
140.19 | |
CHEBI:6824 | |
99% |
Chemical Identifiers
100-97-0 | |
140.19 | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
CHEBI:6824 |
C6H12N4 | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
4101 | |
C1N2CN3CN1CN(C2)C3 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Flammable solid.
GHS P Statement
Wear protective gloves/protective clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 202-905-8
RUO – Research Use Only