HF Scientific™ ProCal™ Turbidity Standard Reagents Kit for Hach™ 2100Q Portable Turbidimeter
Standards not harmful to the environment or to worker safety
Supplier: HF Scientific™ HF52530
Description
- Pre-diluted
- Do not include formazin
- Preservative used enhances stability
- Biodegradable, safe and non-toxic
- Ready and easy to use
- Factory certified by HF scientific
- Custom NTU produced
- Available in different size bottles
Specifications
Primary Turbidity Standard | |
20 NTU | |
8.45 oz. |
Hach 2100Q Portable Turbidimeter | |
EPA | |
250 mL |