missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HF Scientific™ ProCal™ Turbidity Standard Reagents Kit for Hach™ 2100Q Portable Turbidimeter
Standards not harmful to the environment or to worker safety
$273.47 - $283.15
Specifications
For Use With (Equipment) | Hach 2100Q Portable Turbidimeter |
---|---|
Certifications/Compliance | EPA |
Catalog Number | Mfr. No. | Type | Turbidity | Volume (English) | Volume (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Turbidity | Volume (English) | Volume (Metric) | Price | Quantity | ||
15-393-639
![]() |
HF Scientific™
HF52530 |
Primary Turbidity Standard | 20 NTU | 8.45 oz. | 250 mL |
Each for $273.47
|
|
||
15-393-643
![]() |
HF Scientific™
HF52570 |
Primary Turbidity Standard | 800 NTU | 8.45 oz. | 250 mL |
Each for $283.15
|
|
||
Description
- Pre-diluted
- Do not include formazin
- Preservative used enhances stability
- Biodegradable, safe and non-toxic
- Ready and easy to use
- Factory certified by HF scientific
- Custom NTU produced
- Available in different size bottles
Specifications
Hach 2100Q Portable Turbidimeter | |
EPA |